Gene Transcripts
- Gene Symbol
- Fn3krp
- Entrez Gene ID
- 238024
- Gene Name
- fructosamine 3 kinase related protein
- Gene Aliases
- FN3K-RP
- Chromosome Location
- Chr. 11: 121421373 - 121431288 on Build GRCm38
- UniGene ID
- Mm.117394
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC032265.1 |
- |
6 |
1131 |
|
View Full Product Details