Gene Transcripts
- Gene Symbol
- Epb4.1
- Entrez Gene ID
- 269587
- Gene Name
- erythrocyte protein band 4.1
- Gene Aliases
- 4.1R, AI415518, D4Ertd442e, Elp-1, Elp1, Epb41, mKIAA4056
- Chromosome Location
- Chr. 4: 131923413 - 132075579 on Build GRCm38
- UniGene ID
- Mm.30038
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001128606.1 |
NP_001122078.1
|
9, 10 |
1591 |
|
NM_001128607.1 |
NP_001122079.1
|
9, 10 |
1501 |
|
NM_183428.3 |
NP_906273.3
|
9, 10 |
2853 |
|
GenBank mRNA |
AB370261.1 |
- |
8, 9 |
1376 |
|
AK147266.1 |
- |
4, 5 |
547 |
|
AK147644.1 |
- |
4, 5 |
547 |
|
AK220462.1 |
- |
9, 10 |
1498 |
|
BC068138.1 |
- |
9, 10 |
1461 |
|
BC079875.1 |
- |
9, 10 |
1420 |
6844082 |
CN526385.1 |
- |
3, 4 |
435 |
|
L00919.1 |
- |
10, 11 |
2928 |
|
LK021332.1 |
- |
7, 8 |
746 |
|
LK021333.1 |
- |
7, 8 |
746 |
|
View Full Product Details