Gene Transcripts

Gene Symbol
Nol4
Entrez Gene ID
319211
Gene Name
nucleolar protein 4
Gene Aliases
1700013J13Rik, 4930568N03Rik, Gm1262
Chromosome Location
Chr. 18: 22693152 - 23041675 on Build GRCm38
UniGene ID
Mm.209896
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001161483.1 NP_001154955.1 5 513
NM_199024.2 NP_950189.2 5 2283
GenBank mRNA AK031008.1 - 5 2283
BC056377.1 - 5 506 6850243

View Full Product Details