Gene Transcripts

Gene Symbol
Kera
Entrez Gene ID
16545
Gene Name
keratocan
Gene Aliases
CNA2, SLRR2B
Chromosome Location
Chr. 10: 97606959 - 97613693 on Build GRCm38
UniGene ID
Mm.6228
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008438.3 NP_032464.1 2 303
GenBank mRNA AF022256.1 - 2 307
AK086197.1 - 2 327
BC114980.1 - 2 169 40046884

View Full Product Details