Gene Transcripts

Gene Symbol
Krt4
Entrez Gene ID
16682
Gene Name
keratin 4
Gene Aliases
AW108092, K4, Krt-2.4, Krt2-4
Chromosome Location
Chr. 15: 101918535 - 101924735 on Build GRCm38
UniGene ID
Mm.46425
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008475.2 NP_032501.2 9 1834
GenBank mRNA BC064008.1 - 9 1815
X03491.1 - 9 1883

View Full Product Details