Gene Transcripts

Gene Symbol
Lag3
Entrez Gene ID
16768
Gene Name
lymphocyte-activation gene 3
Gene Aliases
CD223, LAG-3, Ly66
Chromosome Location
Chr. 6: 124904359 - 124912435 on Build GRCm38
UniGene ID
Mm.4528
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008479.2 NP_032505.1 6 1442
GenBank mRNA BC120591.1 - 6 1374
X98113.1 - 6 1444

View Full Product Details