Gene Transcripts

Gene Symbol
Tm4sf1
Entrez Gene ID
17112
Gene Name
transmembrane 4 superfamily member 1
Gene Aliases
L6, M3s1
Chromosome Location
Chr. 3: 57287064 - 57301983 on Build GRCm38
UniGene ID
Mm.856
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008536.3 NP_032562.1 7 1782
GenBank mRNA AA088985.1 - 1 268
AK077326.1 - 5 1285
AK133556.1 - 1283

View Full Product Details