Gene Transcripts
- Gene Symbol
- Ncoa2
- Entrez Gene ID
- 17978
- Gene Name
- nuclear receptor coactivator 2
- Gene Aliases
- D1Ertd433e, GRIP-1, Grip1, KAT13C, NCoA-2, SRC-2, TIF2, bHLHe75
- Chromosome Location
- Chr. 1: 13139105 - 13374691 on Build GRCm38
- UniGene ID
- Mm.2537
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AB219976.1 |
- |
5 |
589 |
|
U39060.1 |
- |
7 |
792 |
|
View Full Product Details