Gene Transcripts

Gene Symbol
Dusp8
Entrez Gene ID
18218
Gene Name
dual specificity phosphatase 8
Gene Aliases
5530400B01Rik, AI593498, Nttp1
Chromosome Location
Chr. 7: 142079487 - 142095880 on Build GRCm38
UniGene ID
Mm.39725
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008748.3 NP_032774.1 3 709
GenBank mRNA BC052705.1 - 3 467
X95518.1 - 2 352

View Full Product Details