Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001271705.1 | NP_001258634.1 | 7 | 548 | |
NM_008827.3 | NP_032853.1 | 7 | 881 | ||
GenBank mRNA | AK042891.1 | - | 7 | 865 | |
AK088943.1 | - | 7 | 864 | ||
BC016567.1 | - | 7 | 853 | 2649577 | |
X80171.1 | - | 7 | 650 | ||
X96793.1 | - | 7 | 848 |