Gene Transcripts

Gene Symbol
Lcp2
Entrez Gene ID
16822
Gene Name
lymphocyte cytosolic protein 2
Gene Aliases
AI323664, BB161688, SLP-76, SLP76, twm
Chromosome Location
Chr. 11: 34047201 - 34092280 on Build GRCm38
UniGene ID
Mm.265350
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010696.3 NP_034826.2 21 2402
GenBank mRNA AK036727.1 - 21 2460
BC006948.1 - 21 2404 3603181

View Full Product Details