Gene Transcripts

Gene Symbol
Lipe
Entrez Gene ID
16890
Gene Name
lipase, hormone sensitive
Gene Aliases
4933403G17Rik, HSL
Chromosome Location
Chr. 7: 25379527 - 25398704 on Build GRCm38
UniGene ID
Mm.158548
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039507.2 NP_001034596.1 2 224
NM_010719.5 NP_034849.2 2 660
GenBank mRNA AK019846.1 - 2 1061
AK029984.1 - 2 663
AK169858.1 - 2 219
BC021642.1 - 2 220 4188567
BC053502.1 - 2 284
U08188.1 - 1 745

View Full Product Details