Gene Transcripts

Gene Symbol
Magoh
Entrez Gene ID
17149
Gene Name
mago-nashi homolog, proliferation-associated (Drosophila)
Gene Aliases
Mago-m, Mos2
Chromosome Location
Chr. 4: 107879755 - 107887424 on Build GRCm38
UniGene ID
Mm.808
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001282737.1 NP_001269666.1 5 492
GenBank mRNA AF007862.1 - 5 479
AF035939.1 - 5 439
AK011512.1 - 5 431
BC018176.1 - 5 430 3587774

View Full Product Details