Gene Transcripts
- Gene Symbol
- Mtl5
- Entrez Gene ID
- 17771
- Gene Name
- metallothionein-like 5, testis-specific (tesmin)
- Gene Aliases
- tesmin
- Chromosome Location
- Chr. 19: 3388857 - 3407823 on Build GRCm38
- UniGene ID
- Mm.209941
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001039657.2 |
NP_001034746.1
|
3, 4 |
767 |
|
NM_001039658.2 |
NP_001034747.1
|
3, 4 |
767 |
|
NM_001286557.1 |
NP_001273486.1
|
4, 5 |
851 |
|
NM_001286558.1 |
NP_001273487.1
|
3, 4 |
782 |
|
GenBank mRNA |
AB057423.1 |
- |
3, 4 |
755 |
|
AF329359.1 |
- |
3, 4 |
792 |
|
AF329360.1 |
- |
4, 5 |
861 |
|
AK015724.1 |
- |
3, 4 |
708 |
|
AK015732.1 |
- |
3, 4 |
740 |
|
AK161511.1 |
- |
3, 4 |
768 |
|
BC024377.1 |
- |
3, 4 |
743 |
4976492 |
U67176.1 |
- |
5, 6 |
784 |
|
U77383.1 |
- |
3, 4 |
404 |
|
View Full Product Details