Gene Transcripts
- Gene Symbol
- Nek2
- Entrez Gene ID
- 18005
- Gene Name
- NIMA (never in mitosis gene a)-related expressed kinase 2
- Gene Aliases
- AA617254, C77054
- Chromosome Location
- Chr. 1: 191820875 - 191833049 on Build GRCm38
- UniGene ID
- Mm.33773
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_010892.3 |
NP_035022.2
|
5, 6 |
946 |
|
GenBank mRNA |
AF007247.1 |
- |
5, 6 |
860 |
|
AF013166.1 |
- |
5, 6 |
830 |
|
AK077627.1 |
- |
5, 6 |
910 |
|
AK147072.1 |
- |
5, 6 |
922 |
|
AK164467.1 |
- |
5, 6 |
889 |
|
AK169330.1 |
- |
6, 7 |
837 |
|
AK170179.1 |
- |
5, 6 |
866 |
|
AK171277.1 |
- |
5, 6 |
866 |
|
AK171387.1 |
- |
5, 6 |
866 |
|
BC010302.1 |
- |
5, 6 |
868 |
3597227 |
BC011316.1 |
- |
5, 6 |
866 |
3674001 |
BC057576.1 |
- |
5, 6 |
886 |
6810914 |
U95610.1 |
- |
5, 6 |
857 |
|
View Full Product Details