Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_010907.2 | NP_035037.2 | 5, 6 | 1020 | |
GenBank mRNA | AK089215.1 | - | 5, 6 | 970 | |
AK151115.1 | - | 5, 6 | 973 | ||
AK151608.1 | - | 5, 6 | 973 | ||
AK161622.1 | - | 5, 6 | 932 | ||
BC046754.1 | - | 5, 6 | 953 | 5693175 | |
U36277.1 | - | 5, 6 | 964 |