Gene Transcripts
- Gene Symbol
- Nfkbib
- Entrez Gene ID
- 18036
- Gene Name
- nuclear factor of kappa light polypeptide gene enhancer in B cells inhibitor, beta
- Gene Aliases
- I(Kappa)B(beta), I-kappa-B-beta, IKB-beta, IKappaBbeta, IkB, IkBb, NF-kappa-BIB, ikB-B
- Chromosome Location
- Chr. 7: 28758251 - 28766639 on Build GRCm38
- UniGene ID
- Mm.220333
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK038631.1 |
- |
7 |
1904 |
|
View Full Product Details