Gene Transcripts

Gene Symbol
Mylpf
Entrez Gene ID
17907
Gene Name
myosin light chain, phosphorylatable, fast skeletal muscle
Gene Aliases
2410014J02Rik, MLC-2, Mlc2
Chromosome Location
Chr. 7: 127208848 - 127214288 on Build GRCm38
UniGene ID
Mm.14526
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016754.5 NP_058034.1 7 693
GenBank mRNA AK003149.1 - 7 598
AK010483.1 - 7 694
BC055869.1 - 7 577 4018694
U77943.1 - 6 540

View Full Product Details