Gene Transcripts

Gene Symbol
RBBP5
Entrez Gene ID
5929
Gene Name
RB binding protein 5, histone lysine methyltransferase complex subunit
Gene Aliases
RBQ3, SWD1
Chromosome Location
Chr. 1: 205086142 - 205122022 on Build GRCh38
UniGene ID
Hs.519230
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001193273.1 NP_001180202.1 13 1566
NM_005057.3 NP_005048.2 13 1622
XM_011509833.2 XP_011508135.1 13 1620
GenBank mRNA AK296978.1 - 13 1520
AK297540.1 - 12 1859
BC053856.1 - 13 1593 5576406
BC075059.2 - 13 1520 30915584
BC075060.2 - 13 1520 30915368
X85134.1 - 13 1588

View Full Product Details