Gene Transcripts
- Gene Symbol
- Evx1
- Entrez Gene ID
- 14028
- Gene Name
- even skipped homeotic gene 1 homolog
- Gene Aliases
- Evx-1
- Chromosome Location
- Chr. 6: 52313498 - 52318389 on Build GRCm38
- UniGene ID
- Mm.389754
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_007966.4 |
NP_031992.1
|
3 |
2395 |
|
GenBank mRNA |
X54239.1 |
- |
3 |
2395 |
|
View Full Product Details