Gene Transcripts
- Gene Symbol
- Flot2
- Entrez Gene ID
- 14252
- Gene Name
- flotillin 2
- Gene Aliases
- AI573412, Esa, reggie-2
- Chromosome Location
- Chr. 11: 78037868 - 78060434 on Build GRCm38
- UniGene ID
- Mm.130227
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001040403.1 |
NP_001035493.1
|
8, 9 |
1037 |
|
NM_001284227.1 |
NP_001271156.1
|
9, 10 |
1121 |
|
NM_001284228.1 |
NP_001271157.1
|
9, 10 |
984 |
|
NM_008028.2 |
NP_032054.1
|
9, 10 |
1128 |
|
GenBank mRNA |
AK004589.1 |
- |
8, 9 |
975 |
|
AK030199.1 |
- |
8, 9 |
1000 |
|
AK149530.1 |
- |
9, 10 |
1059 |
|
AK170557.1 |
- |
8, 9 |
1038 |
|
BC070423.1 |
- |
9, 10 |
1086 |
6403816 |
U07890.1 |
- |
9, 10 |
998 |
|
View Full Product Details