Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001290424.1 | NP_001277353.1 | 5 | 680 | |
NM_008031.3 | NP_032057.2 | 5 | 680 | ||
GenBank mRNA | AF461114.1 | - | 5 | 404 | |
AK046605.1 | - | 5 | 640 | ||
AK053211.1 | - | 2 | 422 | ||
AK053701.1 | - | 5 | 640 | ||
AK053829.1 | - | 5 | 682 | ||
AK080948.1 | - | 5 | 640 | ||
AK140343.1 | - | 5 | 642 | ||
BC079671.1 | - | 5 | 666 | 30532682 | |
L23971.1 | - | 5 | 527 |