Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_008175.4 | NP_032201.2 | 5 | 644 | |
GenBank mRNA | AK013678.1 | - | 5 | 460 | |
AK018744.1 | - | 5 | 485 | ||
AK147932.1 | - | 5 | 460 | ||
AK149570.1 | - | 5 | 463 | ||
AK150125.1 | - | 5 | 459 | ||
AK150205.1 | - | 5 | 460 | ||
AK150338.1 | - | 5 | 449 | ||
AK150420.1 | - | 5 | 448 | ||
AK150717.1 | - | 5 | 460 | ||
AK150746.1 | - | 5 | 461 | ||
AK150881.1 | - | 5 | 461 | ||
AK151007.1 | - | 5 | 460 | ||
AK151162.1 | - | 5 | 461 | ||
AK151343.1 | - | 5 | 460 | ||
AK151713.1 | - | 5 | 460 | ||
AK151756.1 | - | 5 | 460 | ||
AK152048.1 | - | 5 | 460 | ||
AK152087.1 | - | 5 | 460 | ||
AK152118.1 | - | 5 | 449 | ||
AK152213.1 | - | 5 | 449 | ||
AK152705.1 | - | 5 | 460 | ||
AK152944.1 | - | 5 | 449 | ||
AK153183.1 | - | 5 | 449 | ||
AK153269.1 | - | 5 | 460 | ||
AK153289.1 | - | 5 | 460 | ||
AK153418.1 | - | 5 | 460 | ||
AK153470.1 | - | 5 | 460 | ||
AK154492.1 | - | 5 | 460 | ||
AK159397.1 | - | 5 | 449 | ||
AK159428.1 | - | 5 | 449 | ||
AK159557.1 | - | 5 | 460 | ||
AK159809.1 | - | 5 | 460 | ||
AK160022.1 | - | 5 | 449 | ||
AK160033.1 | - | 5 | 460 | ||
AK160074.1 | - | 5 | 449 | ||
AK205747.1 | - | 4 | 371 | ||
BC037047.1 | - | 4 | 441 | ||
BC049943.1 | - | 5 | 446 | ||
BC129849.1 | - | 5 | 444 | 40109630 | |
BC129850.1 | - | 5 | 444 | 40109631 | |
M86736.1 | - | 5 | 442 | ||
X62321.1 | - | 4 | 425 |