Gene Transcripts
- Gene Symbol
- H3f3a
- Entrez Gene ID
- 15078
- Gene Name
- H3 histone, family 3A
- Gene Aliases
- H3.3A
- Chromosome Location
- Chr. 1: 180802568 - 180813603 on Build GRCm38
- UniGene ID
- Mm.138832
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_008210.4 |
NP_032236.1
|
2, 3 |
279 |
|
GenBank mRNA |
AK008405.1 |
- |
1, 2 |
179 |
|
AK037900.1 |
- |
2, 3 |
281 |
|
AK150467.1 |
- |
2, 3 |
239 |
|
AK150468.1 |
- |
2, 3 |
239 |
|
AK150591.1 |
- |
2, 3 |
234 |
|
AK150928.1 |
- |
2, 3 |
234 |
|
AK151634.1 |
- |
2, 3 |
239 |
|
AK152293.1 |
- |
2, 3 |
234 |
|
AK153034.1 |
- |
2, 3 |
238 |
|
AK160635.1 |
- |
2, 3 |
234 |
|
AK160677.1 |
- |
2, 3 |
238 |
|
AK162040.1 |
- |
2, 3 |
237 |
|
AK167879.1 |
- |
2, 3 |
238 |
|
AK168197.1 |
- |
2, 3 |
238 |
|
AK169608.1 |
- |
2, 3 |
234 |
|
AK177127.1 |
- |
1, 2 |
49 |
|
AK178894.1 |
- |
1, 2 |
49 |
|
AK180970.1 |
- |
1, 2 |
49 |
|
AK186827.1 |
- |
1, 2 |
49 |
|
AK189335.1 |
- |
1, 2 |
88 |
|
AK203713.1 |
- |
1, 2 |
48 |
|
AK215513.1 |
- |
1, 2 |
49 |
|
AK216872.1 |
- |
|
370 |
|
AK217933.1 |
- |
1, 2 |
49 |
|
BC002268.1 |
- |
2, 3 |
224 |
3495830 |
BC012687.1 |
- |
2, 3 |
228 |
4224280 |
BC083353.1 |
- |
2, 3 |
219 |
5714859 |
BC088835.1 |
- |
2, 3 |
216 |
6437687 |
BC106177.1 |
- |
2, 3 |
213 |
6511787 |
Z85979.1 |
- |
2, 3 |
227 |
|
- Gene Symbol
- Gm12657
- Entrez Gene ID
- 667250
- Gene Name
- predicted gene 12657
- Gene Aliases
- OTTMUSG00000007855
- Chromosome Location
- Chr. 4: 94593650 - 94600319 on Build GRCm38
- UniGene ID
- Mm.449778
- Species
- Mus musculus
- Gene Symbol
- Gm2099
- Entrez Gene ID
- 100039203
- Gene Name
- predicted gene 2099
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Mm.430746
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK137674.1 |
- |
|
2926 |
|
View Full Product Details