Gene Transcripts

Gene Symbol
Inhbe
Entrez Gene ID
16326
Gene Name
inhibin beta E
Gene Aliases
Chromosome Location
Chr. 10: 127349402 - 127351772 on Build GRCm38
UniGene ID
Mm.3510
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008382.2 NP_032408.2 2 1658
GenBank mRNA BC010404.1 - 2 1499
U96386.1 - 2 1647

View Full Product Details