Gene Transcripts
- Gene Symbol
- Fbln1
- Entrez Gene ID
- 14114
- Gene Name
- fibulin 1
- Gene Aliases
- Chromosome Location
- Chr. 15: 85205944 - 85286294 on Build GRCm38
- UniGene ID
- Mm.297992
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK159646.1 |
- |
17 |
2338 |
|
AK159816.1 |
- |
6 |
828 |
|
X70854.1 |
- |
17 |
2380 |
|
View Full Product Details