Gene Transcripts

Gene Symbol
Fem1b
Entrez Gene ID
14155
Gene Name
feminization 1 homolog b (C. elegans)
Gene Aliases
mKIAA0396
Chromosome Location
Chr. 9: 62791824 - 62811648 on Build GRCm38
UniGene ID
Mm.24069
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010193.4 NP_034323.1 2 835
GenBank mRNA AB022863.1 - 2 745
AF064448.1 - 2 566
AF178633.1 - 2 492
AK032338.1 - 2 812
AK122272.1 - 1 33
AK132692.1 - 2 828
AK145371.1 - 1 198
AK149329.1 - 2 798
AK154060.1 - 2 1123
BC068236.1 - 2 789 30544277

View Full Product Details