Gene Transcripts
- Gene Symbol
- Aktip
- Entrez Gene ID
- 14339
- Gene Name
- thymoma viral proto-oncogene 1 interacting protein
- Gene Aliases
- AL023020, Fif, Ft, Ft1, Fts
- Chromosome Location
- Chr. 8: 91109871 - 91135494 on Build GRCm38
- UniGene ID
- Mm.324588
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC008130.1 |
- |
6 |
556 |
|
X71978.1 |
- |
5 |
575 |
|
Z67963.1 |
- |
5 |
575 |
|
View Full Product Details