Gene Transcripts

Gene Symbol
REC8
Entrez Gene ID
9985
Gene Name
REC8 meiotic recombination protein
Gene Aliases
HR21spB, REC8L1, Rec8p
Chromosome Location
Chr. 14: 24171853 - 24180408 on Build GRCh38
UniGene ID
Hs.419259
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001048205.1 NP_001041670.1 3 665
NM_005132.2 NP_005123.2 4 564
XM_011537421.1 XP_011535723.1 3 837
XM_017021841.1 XP_016877330.1 3 634
GenBank mRNA AF006264.1 - 3 137
AK001435.1 - 4 564
AK291191.1 - 4 696
AL832973.1 - 3 555
BC004159.2 - 3 221 2822680
BC010887.1 - 3 598 3535425
CR457263.1 - 3 137
DB294585.1 - 3 665

View Full Product Details