Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001136066.2 | NP_001129538.1 | 3 | 294 | |
NM_010443.2 | NP_034573.2 | 3 | 310 | ||
GenBank mRNA | AF029874.1 | - | 3 | 307 | |
AF054669.1 | - | 3 | 311 | ||
AF054670.1 | - | 3 | 245 | ||
AK032175.1 | - | 3 | 252 | ||
AK134536.1 | - | 3 | 269 | ||
AK141426.1 | - | 3 | 288 | ||
AK152936.1 | - | 3 | 233 | ||
AK179622.1 | - | 2 | 196 | ||
BC002011.1 | - | 3 | 266 | 3484259 | |
BC055742.1 | - | 1 | 1441 | ||
CA465845.1 | - | 3 | 325 |