Gene Transcripts

Gene Symbol
Hoxa11
Entrez Gene ID
15396
Gene Name
homeobox A11
Gene Aliases
Hox-1.9, Hoxa-11
Chromosome Location
Chr. 6: 52242105 - 52245810 on Build GRCm38
UniGene ID
Mm.26954
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010450.3 NP_034580.1 2 1612
GenBank mRNA AK133481.1 - 2 1612
BC052232.1 - 2 1589
U20370.1 - 2 1568

View Full Product Details