Gene Transcripts

Gene Symbol
Hoxc8
Entrez Gene ID
15426
Gene Name
homeobox C8
Gene Aliases
D130011F21Rik, Hox-3.1
Chromosome Location
Chr. 15: 102990539 - 102994254 on Build GRCm38
UniGene ID
Mm.6167
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010466.2 NP_034596.1 2 1198
GenBank mRNA AK051173.1 - 2 654
AK079948.1 - 2 787
AK083688.1 - 1 1196
X07439.1 - 2 1197

View Full Product Details