Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_010511.2 | NP_034641.1 | 7 | 1667 | |
GenBank mRNA | AF128216.1 | - | 7 | 1618 | |
AF128217.1 | - | 7 | 1618 | ||
AK087525.1 | - | 7 | 1657 | ||
AK150503.1 | - | 7 | 1616 | ||
AK151079.1 | - | 7 | 1621 | ||
AK152085.1 | - | 7 | 1616 | ||
AK152384.1 | - | 7 | 1616 | ||
AK153084.1 | - | 7 | 1616 | ||
AK153347.1 | - | 7 | 1619 | ||
AK159359.1 | - | 7 | 1621 | ||
AK159660.1 | - | 7 | 1561 | ||
AK159839.1 | - | 7 | 1561 | ||
AK169593.1 | - | 7 | 1636 | ||
AY157836.1 | - | 7 | 1618 | ||
AY157837.1 | - | 7 | 1618 | ||
BC132599.1 | - | 7 | 1568 | 40130876 | |
BC138617.1 | - | 7 | 1568 | 8861637 | |
J05265.1 | - | 7 | 1644 | ||
M25764.1 | - | 7 | 1569 | ||
M26711.1 | - | 7 | 1644 | ||
M28233.1 | - | 7 | 1640 | ||
M28995.1 | - | 7 | 1545 |