Gene Transcripts

Gene Symbol
Igf2r
Entrez Gene ID
16004
Gene Name
insulin-like growth factor 2 receptor
Gene Aliases
AI661837, CD222, CI-MPR, M6P/IGF2R, Mpr300
Chromosome Location
Chr. 17: 12682406 - 12769706 on Build GRCm38
UniGene ID
Mm.26553
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010515.2 NP_034645.2 4, 5 668
GenBank mRNA BC150817.1 - 4, 5 579
BC171973.1 - 4, 5 579 9054259
U04710.1 - 4, 5 624

View Full Product Details