Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001039701.3 | NP_001034790.1 | 2 | 225 | |
NM_001159562.1 | NP_001153034.1 | 3 | 154 | ||
NM_031167.5 | NP_112444.1 | 3 | 209 | ||
GenBank mRNA | AF001795.1 | - | 3 | 210 | |
AK076296.1 | - | 3 | 210 | ||
AK089222.1 | - | 2 | 180 | ||
AK154601.1 | - | 2 | 180 | ||
AK155263.1 | - | 2 | 180 | ||
AK155641.1 | - | 2 | 226 | ||
AK155756.1 | - | 2 | 180 | ||
AK155798.1 | - | 2 | 180 | ||
AK155846.1 | - | 2 | 180 | ||
AK155888.1 | - | 2 | 222 | ||
AK155908.1 | - | 2 | 180 | ||
AK156034.1 | - | 2 | 180 | ||
AK157157.1 | - | 2 | 218 | ||
AK170378.1 | - | 3 | 215 | ||
AK171037.1 | - | 3 | 158 | ||
AK171385.1 | - | 2 | 213 | ||
BC042532.1 | - | 3 | 186 | 4486098 | |
BY206809.1 | - | 3 | 155 | ||
M57525.1 | - | 2 | 158 | ||
M64404.1 | - | 2 | 240 | ||
M74294.1 | - | 2 | 191 | ||
S64082.1 | - | 2 | 77 |