Gene Transcripts

Gene Symbol
Arf2
Entrez Gene ID
11841
Gene Name
ADP-ribosylation factor 2
Gene Aliases
Chromosome Location
Chr. 11: 103966718 - 103985337 on Build GRCm38
UniGene ID
Mm.5061
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK031259.1 - 2 170
AK042977.1 - 2 210
AK053123.1 - 2 628
AK077591.1 - 2 183
AK167133.1 - 2 180
AK167601.1 - 2 184
AK167653.1 - 2 183
AK169094.1 - 2 180
AK169412.1 - 2 180
BC010487.1 - 2 463 3483748
BY129623.1 - 2 222
D87899.1 - 2 175

View Full Product Details