Gene Transcripts
- Gene Symbol
- Arf5
- Entrez Gene ID
- 11844
- Gene Name
- ADP-ribosylation factor 5
- Gene Aliases
- Chromosome Location
- Chr. 6: 28423640 - 28426499 on Build GRCm38
- UniGene ID
- Mm.389075
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_007480.1 |
NP_031506.1
|
6 |
861 |
|
GenBank mRNA |
D87902.1 |
- |
6 |
861 |
|
View Full Product Details