Gene Transcripts

Gene Symbol
Bpgm
Entrez Gene ID
12183
Gene Name
2,3-bisphosphoglycerate mutase
Gene Aliases
AI323730, AL022789, C86192
Chromosome Location
Chr. 6: 34476254 - 34505610 on Build GRCm38
UniGene ID
Mm.282863
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007563.4 NP_031589.1 2 216
GenBank mRNA AK006315.1 - 2 268
AK037239.1 - 2 217
AK043412.1 - 2 254
AK078119.1 - 2 217
AK171685.1 - 2 217
AK211952.1 - 2 177
BC004589.1 - 2 241 3587820
X13586.1 - 1 109

View Full Product Details