Gene Transcripts

Gene Symbol
C4bp
Entrez Gene ID
12269
Gene Name
complement component 4 binding protein
Gene Aliases
AI195242, C4bpa
Chromosome Location
Chr. 1: 130635921 - 130661618 on Build GRCm38
UniGene ID
Mm.306720
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007576.3 NP_031602.3 8, 9 1265
GenBank mRNA AB094485.1 - 1246
AB094486.1 - 8, 9 1181
AK136735.1 - 8, 9 1179
AK149468.1 - 8, 9 1272
AK149477.1 - 8, 9 1182
AK162345.1 - 8, 9 1177
BC012257.1 - 8, 9 1237 4163203
M17122.1 - 8, 9 1279

View Full Product Details