Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_007737.2 | NP_031763.2 | 2 | 674 | |
GenBank mRNA | AK084285.1 | - | 2 | 456 | |
AK084803.1 | - | 2 | 311 | ||
AK132413.1 | - | 2 | 456 | ||
AK139130.1 | - | 2 | 249 | ||
AK147220.1 | - | 2 | 456 | ||
AK147328.1 | - | 2 | 675 | ||
AK151929.1 | - | 2 | 458 | ||
AK160008.1 | - | 2 | 248 | ||
BC055077.1 | - | 2 | 425 | 30059991 | |
L02918.1 | - | 2 | 306 |