Gene Transcripts

Gene Symbol
Crkl
Entrez Gene ID
12929
Gene Name
v-crk sarcoma virus CT10 oncogene homolog (avian)-like
Gene Aliases
1110025F07Rik, AA589403, AI325100, Crkol
Chromosome Location
Chr. 16: 17451985 - 17487440 on Build GRCm38
UniGene ID
Mm.21048
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001277231.1 NP_001264160.1 2 3142
NM_007764.5 NP_031790.2 3 3608
GenBank mRNA AK048641.1 - 3 3606
AK087977.1 - 1 464

View Full Product Details