Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001301639.1 | NP_001288568.1 | 2 | 1311 | |
NM_001301641.1 | NP_001288570.1 | 2 | 1098 | ||
NM_009688.3 | NP_033818.2 | 3 | 1183 | ||
GenBank mRNA | BC138528.1 | - | 1 | 1106 | |
BC145861.1 | - | 1 | 1106 | 40131164 | |
U36842.1 | - | 2 | 1015 | ||
U88990.1 | - | 1 | 1475 |