Gene Transcripts

Gene Symbol
Neurog2
Entrez Gene ID
11924
Gene Name
neurogenin 2
Gene Aliases
Atoh4, Math4A, bHLHa8, ngn-2, ngn2
Chromosome Location
Chr. 3: 127633145 - 127635631 on Build GRCm38
UniGene ID
Mm.42017
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009718.2 NP_033848.1 2 1261
GenBank mRNA AK143190.1 - 2 1269
BC055743.1 - 2 1261 6808442
Y07621.1 - 2 1031

View Full Product Details