Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_009849.2 | NP_033979.2 | 3 | 358 | |
GenBank mRNA | AF042811.1 | - | 3 | 282 | |
AK002553.1 | - | 3 | 330 | ||
AK172281.1 | - | 3 | 306 | ||
AY376711.1 | - | 3 | 291 | ||
BC011241.1 | - | 3 | 290 | 3582550 | |
U91511.1 | - | 3 | 325 |