Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_009945.3 | NP_034075.2 | 4 | 477 | |
GenBank mRNA | AF037371.1 | - | 4 | 351 | |
AK007875.1 | - | 4 | 354 | ||
AK011269.1 | - | 4 | 337 | ||
AK019210.1 | - | 4 | 332 | ||
AK153173.1 | - | 4 | 354 | ||
BC010979.1 | - | 4 | 322 | 4217409 | |
CN670202.1 | - | 4 | 340 | ||
CN838002.1 | - | 4 | 480 | ||
X58486.1 | - | 4 | 309 |