Gene Transcripts

Gene Symbol
Cry2
Entrez Gene ID
12953
Gene Name
cryptochrome 2 (photolyase-like)
Gene Aliases
AV006279, D130054K12Rik
Chromosome Location
Chr. 2: 92403646 - 92434069 on Build GRCm38
UniGene ID
Mm.254181
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009963.4 NP_034093.1 1, 2 258
GenBank mRNA AB003433.1 - 1, 2 143
AF156987.1 - 1, 2 226
AK037778.1 - 1, 2 226
AK041696.1 - 1, 2 226
AK133781.1 - 1, 2 233
AK172994.1 - 1, 2 190
BC054794.1 - 1, 2 214 6834104
BC066799.1 - 1, 2 217 30103169
CJ164467.1 - 1, 2 258

View Full Product Details