Gene Transcripts

Gene Symbol
Csn1s2b
Entrez Gene ID
12992
Gene Name
casein alpha s2-like B
Gene Aliases
AW987150, Csnd, Csne
Chromosome Location
Chr. 5: 87808121 - 87824421 on Build GRCm38
UniGene ID
Mm.153308
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001301334.1 NP_001288263.1 7, 8 255
NM_001301335.1 NP_001288264.1 6, 7 222
NM_009973.3 NP_034103.2 7, 8 255
GenBank mRNA AK021337.1 - 7, 8 251
AK052780.1 - 7, 8 253
AK085744.1 - 7, 8 255
AK085747.1 - 7, 8 253
AK164763.1 - 7, 8 253
AK164767.1 - 7, 8 255
AK164768.1 - 7, 8 253
AK164795.1 - 7, 8 251
BC002084.1 - 7, 8 236 3489886
J00379.1 - 6, 7 221
V00740.1 - 6, 7 221

View Full Product Details