Gene Transcripts

Gene Symbol
Dusp2
Entrez Gene ID
13537
Gene Name
dual specificity phosphatase 2
Gene Aliases
PAC1
Chromosome Location
Chr. 2: 127336159 - 127338377 on Build GRCm38
UniGene ID
Mm.4729
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010090.2 NP_034220.2 4 1583
GenBank mRNA AK088059.1 - 4 1579
AK134067.1 - 4 1584
BC048696.1 - 4 1569 6511513

View Full Product Details