Gene Transcripts

Gene Symbol
Gdf2
Entrez Gene ID
12165
Gene Name
growth differentiation factor 2
Gene Aliases
Bmp9
Chromosome Location
Chr. 14: 33941039 - 33947198 on Build GRCm38
UniGene ID
Mm.422844
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019506.4 NP_062379.3 2 2820
GenBank mRNA AF156890.1 - 2 2820
AK147029.1 - 2 2821

View Full Product Details