Gene Transcripts

Gene Symbol
Adra2c
Entrez Gene ID
11553
Gene Name
adrenergic receptor, alpha 2c
Gene Aliases
Adra-2c, [a]2C, alpha2-C4, alpha2C
Chromosome Location
Chr. 5: 35278566 - 35281763 on Build GRCm38
UniGene ID
Mm.407152
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007418.3 NP_031444.2 1 2663

View Full Product Details